Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Bromoacetyl)thiophene 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR23196-1G OR23196-5G OR23196-10G
77407-174EA 21.4 USD InStock
77407-174 77407-176 77407-172
2-(2-Bromoacetyl)thiophene 97%
2-(2-Bromoacetyl)thiophene


Formula: C₆H₅BrOS
Melting Pt: 29…33 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD02677721
CAS Number: 10531-41-6
EINECS: 206-994-4
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty