Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-(Hexafluoroisopropylidene)diphenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1229-100G PC1229-1KG PC1229-250G
77413-092EA 36.68 USD InStock
77413-092 77413-094 77413-096
4,4'-(Hexafluoroisopropylidene)diphenol 98%
4,4'-(Hexafluoroisopropylidene)diphenol


Formula: C₁₅H₁₀F₆O₂
Boiling Pt: 350…400 °C
Melting Pt: 160…163 °C
Density: 1.53 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000439
CAS Number: 1478-61-1
EINECS: 216-036-7
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty