Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Diethyl (2-bromoethyl)phosphonate 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR10692-100G OR10692-25G
77402-886EA 399.38 USD InStock
77402-886 77402-888
Diethyl (2-bromoethyl)phosphonate 97%
Diethyl (2-bromoethyl)phosphonate


Formula: C₆H₁₄BrO₃P
MW: 245.06 g/mol
Boiling Pt: 98…100 °C/1 mmHg
Density: 1.348 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000239
CAS Number: 5324-30-1
EINECS: 226-193-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty