Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentafluorophenyl 4-iodobenzenesulfonate

Supplier: APOLLO SCIENTIFIC
Warning

PC10317-1G PC10317-5G
77412-806EA 61.12 USD InStock
77412-806 77412-808
Pentafluorophenyl 4-iodobenzenesulfonate
Pentafluorophenyl 4-iodobenzenesulphonate


Formula: C₁₂H₄F₅IO₃S
Melting Pt: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD05975132
CAS Number: 885950-39-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty