Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydroxy-o-toluic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR11194-100G OR11194-25G
77403-840EA 55.02 USD InStock
77403-840 77403-842
3-Hydroxy-o-toluic acid 99%
3-Hydroxy-o-toluic acid


Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00671541
CAS Number: 603-80-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty