Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-1-methylpiperidine 92%

Supplier: APOLLO SCIENTIFIC
Danger

OR6512-25G OR6512-5G
77410-848EA 32.6 USD InStock
77410-848 77410-850
4-Amino-1-methylpiperidine 92%
4-Amino-1-methylpiperidine


Formula: C₆H₁₄N₂
Boiling Pt: 83 °C/40 mmHg
Density: 0.91 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD01570549
CAS Number: 41838-46-4
UN: 1760
ADR: 8,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty