Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethoxybenzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR11305-1G OR11305-25G OR11305-5G
77403-882EA 21.26 USD InStock
77403-882 77403-884 77403-886
3,5-Dimethoxybenzyl bromide 97%
3,5-Dimethoxybenzyl bromide


Formula: C₉H₁₁BrO₂
Melting Pt: 69…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01321368
CAS Number: 877-88-3
UN: 3261
ADR: 8,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty