Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Difluoro-3-methoxybenzylamine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC303023-25G PC303023-5G
77414-328EA 572.58 USD InStock
77414-328 77414-330
2,6-Difluoro-3-methoxybenzylamine 98%
2,6-Difluoro-3-methoxybenzylamine


Formula: C₈H₉F₂NO
Storage Temperature: Ambient
MDL Number: MFCD04115905
CAS Number: 886498-50-6
UN: 2735
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty