Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-4-(trifluoromethoxy)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0431-1G PC0431-5G
77412-564EA 137 USD InStock
77412-564 77412-566
2-Methyl-4-(trifluoromethoxy)benzyl bromide 97%
2-Methyl-4-(trifluoromethoxy)benzyl bromide


Formula: C₉H₈BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD01631547
CAS Number: 261951-95-5
UN: 1760
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty