Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Lithium hexafluoroantimonate 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC4991-1G PC4991-5G
77415-982EA 41.5 USD InStock
77415-982 77415-986
Lithium hexafluoroantimonate 97%
Lithium hexafluoroantimonate
Useful as a photoinitiator for production of polymers:J. V. Crivello,(GE), US Patent, 4, 136, 102, 1979.


Formula: F₆LiSb
MW: 242.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00042319
CAS Number: 18424-17-4
UN: 1549
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty