Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl acetopyruvate 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR27077-25G OR27077-5G
77407-888EA 68.26 USD InStock
77407-888 77407-890
Ethyl acetopyruvate 97%
Ethyl acetopyruvate


Formula: C₇H₁₀O₄
Boiling Pt: 101…103 °C/12 mmHg
Density: 1.126 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00009124
CAS Number: 615-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty