Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,4,5-Trichlorophenoxy)propionic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR315333-25G OR315333-5G
77409-060EA 111.06 USD InStock
77409-060 77409-062
2-(2,4,5-Trichlorophenoxy)propionic acid
2-(2,4,5-Trichlorophenoxy)propionic acid


Formula: C₉H₇Cl₃O₃
Melting Pt: 175…177 °C
Storage Temperature: Ambient
MDL Number: MFCD00002646
CAS Number: 93-72-1
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty