Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5'-Bromo-2'-hydroxypropiophenone

Supplier: APOLLO SCIENTIFIC
Danger

OR22402-1G OR22402-5G
77406-878EA 23 USD InStock
77406-878 77406-880
5'-Bromo-2'-hydroxypropiophenone
5'-Bromo-2'-hydroxypropiophenone


Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00218577
CAS Number: 17764-93-1
UN: 2811
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty