Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxy-3-methylphenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10528-100G OR10528-25G OR10528-5G
77402-766EA 564.42 USD InStock
77402-766 77402-768 77402-770
4-Methoxy-3-methylphenylboronic acid 98%
4-Methoxy-3-methylphenylboronic acid


Formula: C₈H₁₁BO₃
MW: 165.99 g/mol
Melting Pt: 194…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179464
CAS Number: 175883-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty