Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Acetyl iodide 95%

Supplier: APOLLO SCIENTIFIC

OR938283-1G OR938283-5G OR938283-10G
77412-034EA 136.52 USD InStock
77412-034 77412-036 77412-032
Acetyl iodide 95%
Acetyl iodide


Formula: C₂H₃IO
Storage Temperature: Ambient
MDL Number: MFCD00039399
CAS Number: 507-02-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty