Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-4-nitrobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC57031-10G PC57031-1G PC57031-5G
77416-946EA 223.12 USD InStock
77416-946 77416-948 77416-950
2-Fluoro-4-nitrobenzaldehyde 97%
2-Fluoro-4-nitrobenzaldehyde


Formula: C₇H₄FNO₃
Melting Pt: 98…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00968941
CAS Number: 157701-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty