Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dimethyl-1H-pyrazole-4-boronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR18577-100MG OR18577-1G OR18577-250MG
77406-346EA 73.36 USD InStock
77406-346 77406-348 77406-350
1,5-Dimethyl-1H-pyrazole-4-boronic acid 98%
1,5-Dimethyl-1H-pyrazole-4-boronic acid


Formula: C₅H₉BN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD13195678
CAS Number: 1204333-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty