Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexylboronic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR13116-100G OR13116-25G OR13116-5G
77404-522EA 241.46 USD InStock
77404-522 77404-524 77404-526
Cyclohexylboronic acid 97%
Cyclohexylboronic acid


Formula: C₆H₁₃BO₂
MW: 127.9772 g/mol
Melting Pt: 122…123 °C
Storage Temperature: Freezer
MDL Number: MFCD01311824
CAS Number: 4441-56-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty