Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Acetyl-4-chloro-1H-indazole 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR59366-1G OR59366-25G OR59366-5G
77410-570EA 103.92 USD InStock
77410-570 77410-572 77410-574
1-Acetyl-4-chloro-1H-indazole 98%
1-Acetyl-4-chloro-1H-indazole


Formula: C₉H₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09878372
CAS Number: 145439-15-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty