Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-bromo-6-chloropyridazine 98%

Supplier: APOLLO SCIENTIFIC

OR480266-25G OR480266-5G
77410-004EA 109 USD InStock
77410-004 77410-006
3-Amino-4-bromo-6-chloropyridazine 98%
3-Amino-4-bromo-6-chloropyridazine


Formula: C₄H₃BrClN₃
Storage Temperature: Ambient
MDL Number: MFCD11520891
CAS Number: 446273-59-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty