Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-chloro-1H-indazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR17924-1G OR17924-250MG
77406-156EA 37 USD InStock
77406-156 77406-158 77406-160
5-Bromo-3-chloro-1H-indazole 97%
5-Bromo-3-chloro-1H-indazole


Formula: C₇H₄BrClN₂
Storage Temperature: Ambient
MDL Number: MFCD06738284
CAS Number: 36760-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty