Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Tributylstannyl)pyrimidine

Supplier: APOLLO SCIENTIFIC
Danger

OR52038-1G OR52038-5G OR52038-10G
77410-194EA 79.46 USD InStock
77410-194 77410-196 77410-192
2-(Tributylstannyl)pyrimidine
2-(Tributylstannyl)pyrimidine


Formula: C₁₆H₃₀N₂Sn
Density: 1.164 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01319085
CAS Number: 153435-63-3
UN: 2788
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty