Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxytetrahydropyran 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR14305-100G OR14305-25G OR14305-5G
77404-894EA 110.04 USD InStock
77404-894 77404-896 77404-898
4-Hydroxytetrahydropyran 97%
4-Hydroxytetrahydropyran


Formula: C₅H₁₀O₂
Boiling Pt: 87 °C/15 mmHg
Density: 1.071 g/cm³ (25 °C)
Flash Pt: 88 °C
Storage Temperature: Ambient
MDL Number: MFCD00006633
CAS Number: 2081-44-9
EINECS: 218-210-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty