Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxypyridine-2-carbonitrile 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR9544-1G OR9544-5G
77412-184EA 75.4 USD InStock
77412-184 77412-186
4-Methoxypyridine-2-carbonitrile 95%
4-Methoxypyridine-2-carbonitrile


Formula: C₇H₆N₂O
Melting Pt: 118…121 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08062857
CAS Number: 36057-44-0
UN: 3439
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty