Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethyl-4-(tributylstannyl)isoxazole 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR15592-100MG OR15592-1G OR15592-250MG
77405-478EA 69.28 USD InStock
77405-478 77405-480 77405-482
3,5-Dimethyl-4-(tributylstannyl)isoxazole 95%
3,5-Dimethyl-4-(tributylstannyl)isoxazole


Formula: C₁₇H₃₃NOSn
Storage Temperature: Ambient
MDL Number: MFCD01318973
CAS Number: 136295-80-2
UN: 2788
ADR: 6.1 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty