Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-2-methylpiperazine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR16016-250MG OR16016-25G OR16016-5G
77405-610EA 20.4 USD InStock
77405-610 77405-612 77405-614
1-Boc-2-methylpiperazine 95%
1-Boc-2-methylpiperazine


Formula: C₁₀H₂₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD02179168
CAS Number: 120737-78-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty