Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1H-Imidazol-4-yl)methanol hydrochloride 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR2014-25G OR2014-5G OR2014-1G
77406-554EA 137.54 USD InStock
77406-554 77406-556 77406-552
(1H-Imidazol-4-yl)methanol hydrochloride 98+%
(1H-Imidazol-4-yl)methanol hydrochloride


Formula: C₄H₇ClN₂O
Melting Pt: 108…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012697
CAS Number: 32673-41-9
EINECS: 251-150-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty