Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Biphenyl-4-sulfonyl chloride 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR11127-10G OR11127-25G OR11127-5G
77403-700EA 25 USD InStock
77403-700 77403-702 77403-706
Biphenyl-4-sulfonyl chloride 95%
Biphenyl-4-sulphonyl chloride


Formula: C₁₂H₉ClO₂S
MW: 252.72 g/mol
Boiling Pt: 200…205 °C
Melting Pt: 112…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00078658
CAS Number: 1623-93-4
EINECS: 216-609-1
UN: 3261
ADR: 8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty