Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2,4-difluoroaniline 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC9925-5G PC9925-25G
77418-794EA 21.26 USD InStock
77418-794 77418-792
5-Bromo-2,4-difluoroaniline 98%
5-Bromo-2,4-difluoroaniline


Formula: C₆H₄BrF₂N
Melting Pt: 25…29 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03426368
CAS Number: 452-92-6
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty