Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR950059-1G OR950059-25G OR950059-5G
77412-140EA 21.4 USD InStock
77412-140 77412-142 77412-144
tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate 97%
tert-Butyl-3-endo-3-hydroxy-8-azabicyclo[3.2.1]octane-8-carboxylate


Formula: C₁₂H₂₁NO₃
Melting Pt: 141…142.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD22409620
CAS Number: 143557-91-9
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty