Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Piperidin-1-yl)benzaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR1647-5G OR1647-10G
77405-718EA 117.16 USD InStock
77405-718 77405-710
2-(Piperidin-1-yl)benzaldehyde 95%
2-(Piperidin-1-yl)benzaldehyde


Formula: C₁₂H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD03419311
CAS Number: 34595-26-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty