Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Methyl-1H-indole-3-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR10175-1G OR10175-25G OR10175-5G
77402-520EA 21.26 USD InStock
77402-520 77402-522 77402-524
2-Methyl-1H-indole-3-carbaldehyde
2-Methyl-1H-indole-3-carbaldehyde


Formula: C₁₀H₉NO
MW: 159.1846 g/mol
Melting Pt: 200…205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012077
CAS Number: 5416-80-8
EINECS: 226-512-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty