Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl azetidin-3-ylcarbamate

Supplier: APOLLO SCIENTIFIC
Warning

OR6153-10G OR6153-1G OR6153-5G
77410-768EA 220.38 USD InStock
77410-768 77410-770 77410-772
tert-Butyl azetidin-3-ylcarbamate
tert-Butyl azetidin-3-ylcarbamate


Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01861759
CAS Number: 91188-13-5
UN: 3077
ADR: 9+EHS III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty