Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Piperazine-2-carboxylic acid dihydrochloride 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR15638-10G OR15638-1G OR15638-5G
77405-492EA 78.44 USD InStock
77405-492 77405-494 77405-496
(S)-Piperazine-2-carboxylic acid dihydrochloride 95%
(S)-Piperazine-2-carboxylic acid dihydrochloride


Formula: C₅H₁₂Cl₂N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD01632104
CAS Number: 158663-69-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty