Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-methylbutanoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR01641-1G OR01641-25G OR01641-5G
77401-788EA 21.4 USD InStock
77401-788 77401-790 77401-792
3-Amino-3-methylbutanoic acid
3-Amino-3-methylbutanoic acid


Formula: C5H11NO2
Melting Pt: 232…233 °C
Storage Temperature: Ambient
MDL Number: MFCD06245495
CAS Number: 625-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty