Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Hydroxymethyl)phenoxyacetic acid 97%

Supplier: APOLLO SCIENTIFIC

OR16415-25G OR16415-5G
77405-682EA 38.72 USD InStock
77405-682 77405-684
4-(Hydroxymethyl)phenoxyacetic acid 97%
4-(Hydroxymethyl)phenoxyacetic acid


Formula: C₉H₁₀O₄
MW: 182.2 g/mol
Melting Pt: 107…114 °C
Storage Temperature: Freezer
MDL Number: MFCD00057827
CAS Number: 68858-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty