Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Difluoromethyl)trimethylsilane 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC408038-25G PC408038-5G
77415-086EA 206.82 USD InStock
77415-086 77415-088
(Difluoromethyl)trimethylsilane 98%
(Difluoromethyl)trimethylsilane


Formula: C₄H₁₀F₂Si
MW: 124.21 g/mol
Boiling Pt: 65…66 °C
Density: 0.91 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD19381658
CAS Number: 65864-64-4
UN: 1993
ADR: 3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty