Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,5,6-Tetrafluoro-p-xylene 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC0582-1G PC0582-250MG
77412-646EA 34 USD InStock
77412-646 77412-648 77412-650
2,3,5,6-Tetrafluoro-p-xylene 98%
2,3,5,6-Tetrafluoro-p-xylene


Formula: C₈H₆F₄
Boiling Pt: 143…144 °C
Melting Pt: 35…37 °C
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00012231
CAS Number: 703-87-7
UN: 1325
ADR: 4.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty