Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95+%

Supplier: APOLLO SCIENTIFIC
Danger

OR5367-10G OR5367-1G OR5367-5G
77410-322EA 28.5 USD InStock
77410-322 77410-324 77410-326
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95+%
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite


Formula: C₁₅H₃₂N₃OP
Boiling Pt: 100 °C/0.5 mmHg
Density: 0.949 g/cm³ (25 °C)
Flash Pt: 60 °C
Storage Temperature: Freezer
MDL Number: MFCD00012213
CAS Number: 102691-36-1
UN: 1993
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty