Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Dimethylamino)acrylonitrile 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR7503-1G OR7503-25G OR7503-5G
77411-228EA 30.56 USD InStock
77411-228 77411-230 77411-232
3-(Dimethylamino)acrylonitrile 97+%
3-(Dimethylamino)acrylonitrile


Formula: C₅H₈N₂
Boiling Pt: 76…80 °C/0.3 mmHg
Density: 0.95 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00001929
CAS Number: 2407-68-3
EINECS: 219-305-7
UN: 3276
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty