Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxychrysoidine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

OR10870-10G OR10870-1G OR10870-5G
77402-958EA 212.94 USD InStock
77402-958 77402-960 77402-962
4-Ethoxychrysoidine hydrochloride
4-Ethoxychrysoidine hydrochloride


Formula: C₁₄H₁₆N₄O
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00042023
CAS Number: 2313-87-3
EINECS: 219-010-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty