Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibromoisocyanuric acid

Supplier: APOLLO SCIENTIFIC
Danger

OR16500-1G OR16500-25G OR16500-5G
77405-726EA 21.4 USD InStock
77405-726 77405-728 77405-730
Dibromoisocyanuric acid
Dibromoisocyanuric acid


Formula: C₃HBr₂N₃O₃
MW: 286.87 g/mol
Melting Pt: 309 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00463941
CAS Number: 15114-43-9
UN: 3085
ADR: 5.1+8 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty