Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1,1,2,2-tetrafluoroethyl ether

Supplier: APOLLO SCIENTIFIC
Danger

PC3265-100G PC3265-25G
77414-622EA 256.74 USD InStock
77414-622 77414-624
Ethyl-1,1,2,2-tetrafluoroethyl ether
Ethyl-1,1,2,2-tetrafluoroethyl ether


Formula: C₄H₆F₄O
Boiling Pt: 57 °C
Density: 1.198 g/cm³ (25 °C)
Flash Pt: –15 °C
Storage Temperature: Ambient
MDL Number: MFCD00069162
CAS Number: 512-51-6
UN: 3271
ADR: 3,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty