Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-2-fluoropropionate 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC9352-100G PC9352-25G PC9352-5G
77418-662EA 94.76 USD InStock
77418-662 77418-664 77418-666
Ethyl-2-fluoropropionate 97%
Ethyl-2-fluoropropionate


Formula: C₅H₉FO₂
Boiling Pt: 121…125 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD01320801
CAS Number: 349-43-9
UN: 3272
ADR: 3,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty