Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium-2-mercaptobenzothiazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR938282-100G OR938282-25G OR938282-500G
77412-026EA 41.78 USD InStock
77412-026 77412-028 77412-030
Sodium-2-mercaptobenzothiazole 97%
Sodium-2-mercaptobenzothiazole


Formula: C₇H₅NS₂
Storage Temperature: Ambient
MDL Number: MFCD00065334
CAS Number: 2492-26-4
UN: 3077
ADR: 9+EHS,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty