Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Diamino-6-[3-(trifluoromethyl)phenyl]-1,3,5-triazine

Supplier: APOLLO SCIENTIFIC
Danger

PC6632-1G PC6632-25G PC6632-5G
77417-462EA 69.36 USD InStock
77417-462 77417-464 77417-466
2,4-Diamino-6-[3-(trifluoromethyl)phenyl]-1,3,5-triazine
2,4-Diamino-6-[3-(trifluoromethyl)phenyl]-1,3,5-triazine


Formula: C₁₀H₈F₃N₅
Boiling Pt: 333…334 °C
Melting Pt: 207…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00268708
CAS Number: 30508-78-2
UN: 2811
ADR: 6.1,II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty