Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid

Supplier: APOLLO SCIENTIFIC

OR60132-1G OR60132-250MG
77410-678EA 55.02 USD InStock
77410-678 77410-680
1-(3-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid
1-(3-Chlorophenyl)-5-oxopyrrolidine-3-carboxylic acid


Formula: C₁₁H₁₀ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD01330895
CAS Number: 92847-41-1
UN: 2811
ADR: 6.1,III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty