Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Difluoromethyl)benzene 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC0683-1G PC0683-25G PC0683-5G
77412-680EA 38.72 USD InStock
77412-680 77412-682 77412-684
(Difluoromethyl)benzene 95%
(Difluoromethyl)benzene


Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 133…139 °C
Density: 1.138 g/cm³ (25 °C)
Flash Pt: 35.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03788488
CAS Number: 455-31-2
UN: 2920
ADR: 8+3 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty