Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC2021-1G PC2021-250MG
77413-486EA 72.34 USD InStock
77413-486 77413-488
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride 97%
5-(4-Chlorophenyl)-2-(trifluoromethyl)furan-3-carbonyl chloride


Formula: C₁₂H₅Cl₂F₃O₂
MW: 309.07 g/mol
Melting Pt: 135…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00275554
CAS Number: 175276-61-6
UN: 1759
ADR: 8 III
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty