Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)-DL-phenylalanine

Supplier: APOLLO SCIENTIFIC
Warning

PC7714M-1G PC7714M-5G PC7714M-10G
77418-134EA 87.26 USD InStock
77418-134 77418-136 77418-132
3-(Trifluoromethyl)-DL-phenylalanine
3-(Trifluoromethyl)-DL-phenylalanine


Formula: C₁₀H₁₀F₃NO₂
MW: 233.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00069074
CAS Number: 63701-37-1
UN: 2811
ADR: 6.1 II
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty